missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) sulfate, ≥99%, Reagent Grade, Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
Description
- ≥95% for general laboratory use.
Specifications
Specifications
| Color | Gray |
| Physical Form | Powder |
| Quantity | 500 g |
| Linear Formula | CuSO4 |
| UN Number | UN3077 |
| Formula Weight | 159.61g/mol |
| Percent Purity | ≥99% |
| Packaging | Glass bottle |
| Vapor Pressure | 7.3mmHg (25°C) |
| Density | 3.603g/mL at 25°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction