missing translation for 'onlineSavingsMsg'
Learn More

Copper (II) Sulfate Pentahydrate (Crystalline (Blue)), Fisher BioReagents

Product Code. 10669163
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10669163 500 g Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10669163 Supplier Fisher BioReagents Supplier No. BP346500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7758-98-7 | CuO4S | 159.602 g/mol

Glass Bottle; 500g

Copper Sulfate is suitable for use in tissue culture systems requiring additives.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Copper (II) Sulfate Pentahydrate
Melting Point 110°C
CAS 7758-98-7, 7758-99-8
Color Blue
pH 3.5 to 4.5
Physical Form Solid
Quantity 500 g
Assay Percent Range 98.0 to 102.0 %
Molecular Formula CuO4S
Merck Index 15, 2643
Synonym copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle
InChI Key ARUVKPQLZAKDPS-UHFFFAOYSA-L
SMILES [O-]S(=O)(=O)[O-].[Cu+2]
IUPAC Name copper;sulfate
Molecular Weight (g/mol) 159.602
PubChem CID 24462
ChEBI CHEBI:23414
Formula Weight 249.68
Percent Purity 98.0 to 102.0%
Identification Pass Test
Packaging Glass Bottle
Vapor Pressure 7.3mmHg at 25°C
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Copper(II) sulfate pentahydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contain: Copper (II) sulfatepentahydrate (1:1:5), Cupric sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.