missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) Sulfate Pentahydrate, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Copper sulfate pentahydrate |
| CAS | 7758-99-8 |
| Molecular Formula | Cu . SO4 . 5[H2O] |
| Quantity | 5 g |
| Synonym | Copper(II) sulphate pentahydrate, Sulfuric acid copper(2+) Salt (1:1) Pentahydrate, Copper Sulfate (CuSO4) Pentahydrate, Copper Sulfate Pentahydrate, Copper(2+) Sulfate Pentahydrate, Copper(II) Sulfate Pentahydrate, Cupric Sulfate Pentahydrate, Sulfuric acid copper(2+) salt (1:1), pentahydrate |
| InChI Formula | InChI=1S/Cu.H2O4S.5H2O/c;1-5(2,3)4;;;;;/h;(H2,1,2,3,4);5*1H2/q+2;;;;;;/p-2 |
| SMILES | [O-]S([O-])(=O)=O.[Cu+2].O.O.O.O.O |
| IUPAC Name | copper;sulfate;pentahydrate |
| Molecular Weight (g/mol) | 249.68 |
| Formula Weight | 248.9342 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction