missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Sulfate Pentahydrate, TRC

Product Code. 30365106
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365106 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30365106 Supplier Toronto Research Chemicals Supplier No. TRCC685370250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Copper sulfate pentahydrate
CAS 7758-99-8
Molecular Formula Cu . SO4 . 5[H2O]
Quantity 250 g
Synonym Copper(II) sulphate pentahydrate, Sulfuric acid copper(2+) Salt (1:1) Pentahydrate, Copper Sulfate (CuSO4) Pentahydrate, Copper Sulfate Pentahydrate, Copper(2+) Sulfate Pentahydrate, Copper(II) Sulfate Pentahydrate, Cupric Sulfate Pentahydrate, Sulfuric acid copper(2+) salt (1:1), pentahydrate
InChI Formula InChI=1S/Cu.H2O4S.5H2O/c;1-5(2,3)4;;;;;/h;(H2,1,2,3,4);5*1H2/q+2;;;;;;/p-2
SMILES [O-]S([O-])(=O)=O.[Cu+2].O.O.O.O.O
IUPAC Name copper;sulfate;pentahydrate
Molecular Weight (g/mol) 249.68
Formula Weight 248.9342
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.