missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Sulfate, TRC

Product Code. 30136363
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136363 10 g 10g
30134727 5 g 5g
30004318 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136363 Supplier Toronto Research Chemicals Supplier No. TRCC72504010G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-98-7
Molecular Formula Cu . O4 S
Molecular Weight (g/mol) 159.61
Synonym Cupric sulfate, Copper(2+) sulfate (1:1) (ACI), Baijunling, Black blue stone, Blue Copper, Blue stone, Blue vitriol, Bluestone, Copper monosulfate, Copper sulfate, Copper sulfate (1:1), Copper sulfate (CuSO4), Copper(2+) sulfate, Copper(II) sulfate, Cuivrol, Cupric sulfate anhydrous, Copper (II) sulphate anhydrous, Cupric sulphate, CuPRiX20, Delcup, EarthTec, Hylinec, Incracide 10A, Incracide E 51, MAC 570, Mastercop, Monocopper sulfate, Reducer Cu, Roman vitriol, Sulfuric acid, copper(2+) salt (1:1), Sulfuric acid, monocopper(1+) salt, Upinorg, XWHGS 005
IUPAC Name copper;sulfate
SMILES [Cu+2].[O-]S(=O)(=O)[O-]

Specifications

Quantity 10 g
InChI Formula InChI=1S/Cu.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2
Formula Weight 158.8813
Chemical Name or Material Copper sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.