missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) tetrafluoroborate, 45% aq. soln.

Product Code. 15473007
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
Product Code. Quantity unitSize
15473007 1 kg 1kg
1 options
This item is not returnable. View return policy

Product Code. 15473007

Brand: Thermo Scientific Alfa Aesar 089666.A1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 38465-60-0 | B2CuF8 | 237.15 g/mol

Copper(II) tetrafluoroborate is used in copper plating and in the production of printed circuits. It serves as an intermediate for homometallic, trinuclear heteroscorpionate complexes applied in studies of electronic and magnetic properties. It is also employed in the promotion of Diels-Alder reactions and decomposition of diazo compounds. Further, it is used as a component of fluxing and plating, as a catalyst and as an active filler in resin bonded abrasives. It is used in flame-retardant production, metal treatment, grain refining agents, electrolytic generation of boron and in the preparation of glazing frits. In addition it functions as an oxidizing agent and reacts with siloxycyclopropanes to generate beta acylalkylcopper(II) species.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 38465-60-0
Color Blue
Physical Form Liquid
Quantity 1 kg
Molecular Formula B2CuF8
MDL Number MFCD00016054
UN Number UN3264
Synonym copper fluoroborate, copper fluoborate, copper ii tetrafluoroborate, copper ditetrafluoroborate, cupric tetrafluoroborate hydrate, copper ii tetrafluoroborate in h2o, cupric fluoborate, copper ii fluoborate, copper 2+ ditetrafluoroborate, copper 2+ tetrafluoroborate 1-
Solubility Information Soluble in water. Slightly soluble in alcohol.
InChI Key HMUNZEYTSRPVBE-UHFFFAOYSA-N
SMILES [Cu++].F[B-](F)(F)F.F[B-](F)(F)F
IUPAC Name copper;ditetrafluoroborate
Molecular Weight (g/mol) 237.15
PubChem CID 170058
Formula Weight 237.15
Concentration or Composition (by Analyte or Components) 45% aq. soln.
Chemical Name or Material Copper (II) tetrafluoroborate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.