missing translation for 'onlineSavingsMsg'
Learn More

Copper Nickel wire, 0.13mm (0.005in) dia

Product Code. 11349378
Change view
Click to view available options
Quantity:
100 m
500 m
Unit Size:
100m
500m
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349378 100 m 100m
11359378 500 m 500m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11349378 Supplier Thermo Scientific Alfa Aesar Supplier No. 043270.H4
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Form: Approx. 0.11g/m | Cu:Ni 55:45 wt%

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Copper Nickle
Color Silver
Appearance Silver-colored wire
Quantity 100 m
Odor Odorless
Form Wire
Molecular Formula Cu:Ni 55:45 wt%
MDL Number MFCD00801098

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.