missing translation for 'onlineSavingsMsg'
Learn More

Copper Nickel wire, 1.6mm (0.064 in.) dia., Thermo Scientific™

Product Code. 11339388
Change view
Click to view available options
Quantity:
100 m
25 m
5 m
Unit Size:
100m
25m
5m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11339388 100 m 100m
11329388 5 m 5m
11319388 25 m 25m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11339388 Supplier Thermo Scientific Alfa Aesar Supplier No. 043273.H4
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Copper Nickel wire
Name Note 1.6mm (0.064 in.) dia.
Avg. Mol. Wt. or Mol. Wt. Range Cu:Ni 55:45 wt%
Quantity 100 m
Weight ≈17.9g/m
Form Wire
MDL Number MFCD00801098
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.