missing translation for 'onlineSavingsMsg'
Learn More

Copper phthalocyanine, dye content ca 95%

Product Code. 10752671 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10752671 25 g Glass bottle 25g
10489940 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10752671 Supplier Thermo Scientific Acros Supplier No. 269150250
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 147-14-8 | C32H16CuN8

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147-14-8
Molecular Formula C32H16CuN8
MDL Number MFCD00010719
Synonym C.I. 74160, Pigment blue 15

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
UV Spectrometry Complies
Merck Index 15, 2505
Color Blue
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in 98% h2so4, practically insoluble in alcohol and hydrocarbons
Quantity 25 g
Physical Form Fine Crystalline Powder
Formula Weight 576.08
Percent Purity ≥94% (ex Cu)
Chemical Name or Material Copper phthalocyanine, dye content ca 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.