missing translation for 'onlineSavingsMsg'
Learn More

Copper powder, -325 mesh, 10% max +325 mesh, 99% (metals basis)

Product Code. 11301509
Change view
Click to view available options
Quantity:
50 g
500 g
2 kg
10 kg
Unit Size:
10kg
2kg
500g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11301509 500 g 500g
11331509 50 g 50g
11311509 2 kg 2kg
11321509 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11301509 Supplier Thermo Scientific Alfa Aesar Supplier No. 013990.36
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7440-50-8 | Cu | 63.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Quantity 500 g
Mesh Size −325 Mesh 10% max +325 Mesh
Odor Odorless
Particle Size APS 8 to 11 Micron (FSSS)
Form Powder
UN Number UN3089
Percent Purity 99%
Assay (metals basis)
Chemical Name or Material Copper powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.