missing translation for 'onlineSavingsMsg'
Learn More

Copper wire, 0.025mm (0.001in) dia, Puratronic™, 99.995% (metals basis)

Product Code. 11361548
Change view
Click to view available options
Quantity:
10 m
50 m
200 m
Unit Size:
10m
200m
50m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361548 50 m 50m
11371548 10 m 10m
11381548 200 m 200m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11361548 Supplier Thermo Scientific Alfa Aesar Supplier No. 010970.H2
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-50-8 | Cu | 63.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Quantity 50 m
Odor Odorless
Weight ≈0.0044g/m
Form Wire
Percent Purity 99.995%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Copper wire, 0.025mm (0.001 in.) dia.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.