missing translation for 'onlineSavingsMsg'
Learn More

Copper, technical, metal wire, about 0.91mm, 20 swg, Fisher Chemical

Product Code. 10000100 Shop All Fisher Chemical Products
Click to view available options
:
500g, Plastic container
This item is not returnable. View return policy

Product Code. 10000100

Brand: Fisher Chemical C/7210/53

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color Brown
CAS 7440-50-8
Melting Point 1083°C
Boiling Point 2595°C
Quantity 500 g
Packaging Plastic container
Form Solid
Molecular Formula Cu
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES [Cu]
IUPAC Name copper
Molecular Weight (g/mol) 63.55
Formula Weight 63.54g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.