missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ APY 29

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Allosteric modulator of IRE1α. Inhibits IRE1α autophosphorylation (IC50 = 280 nM) and activates IRE1α ribonuclease activity.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 332.36 |
| Physical Form | Solid |
| Color | White |
| Purity | >98% |
| Molecular Weight (g/mol) | 332.371 |
| Chemical Name or Material | APY 29 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction