missing translation for 'onlineSavingsMsg'
Learn More

Proteintech CoraLite™594-Phalloidin (red)

Product Code. 17521176 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
300 Tests
Unit Size:
300 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17521176 300 Tests 300 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17521176 Supplier Proteintech Supplier No. PF00003300T
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Phalloidin is a toxin isolated from mushrooms that binds F-Actin in the cytoskeleton.

  • This product is conjugated to our fluorescent CoraLite(R) dye to stain F-actin for labeling, identifying and quantifying actin filaments in immunostaining studies.
  • Compared to many fluorescent dyes, CoraLite(R) dyes have advantages in brightness, light stability and water solubility.
  • Excitation/Emission maxima wavelengths are 593 nm/614 nm

Specifications

For Use With (Application) Immunofluorescence
Quantity 300 Tests
Emission 614 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.