missing translation for 'onlineSavingsMsg'
Learn More

Cordycepin, MedChemExpress

Product Code. 16418928
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16418928 10 mg 10mg
16484797 50 mg 50mg
16473846 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16418928 Supplier Medchem Express Supplier No. HYN0262
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cordycepin (3'-Deoxyadenosine) is a nucleoside derivative and inhibits IL-1β-induced MMP-1 and MMP-3 expression in rheumatoid arthritis synovial fibroblasts (RASFs) in a dose-dependent manner. Cordycepin kills Mycobacterium tuberculosis through hijacking the bacterial adenosine kinase.

Cordycepin (3'-Deoxyadenosine) is a nucleoside derivative and inhibits IL-1β-induced MMP-1 and MMP-3 expression in rheumatoid arthritis synovial fibroblasts (RASFs) in a dose-dependent manner. Cordycepin kills Mycobacterium tuberculosis through hijacking the bacterial adenosine kinase.

Specifications

Chemical Name or Material Cordycepin
CAS 73-03-0
Color White
Purity Grade Notes Research
Molecular Formula C10H13N5O3
Quantity 10 mg
Synonym 3'-Deoxyadenosine
Solubility Information DMSO : 50 mg/mL (199.01 mM; Need ultrasonic) ∣H2O : 2 mg/mL (7.96 mM; ultrasonic and warming and heat to 60°C)
SMILES OC[C@@H]1C[C@H]([C@H](N2C=NC3=C2N=CN=C3N)O1)O
Molecular Weight (g/mol) 251.24
Formula Weight 251.24
Percent Purity 98.64%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.