missing translation for 'onlineSavingsMsg'
Learn More

Cordycepin, TRC

Product Code. 30007080
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007080 100 mg 100mg
30007244 25 mg 25mg
30006937 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007080 Supplier Toronto Research Chemicals Supplier No. TRCC685800100MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-03-0
Molecular Formula C10 H13 N5 O3
Molecular Weight (g/mol) 251.24
Synonym Adenosine, 3'-deoxy-, Cordycepin (6CI,7CI), 3'-Deoxyadenosine, 9-(3-Deoxy-β-D-ribofuranosyl)adenine, 9-Cordyceposidoadenine, 9H-Purin-6-amine, 9-(3-deoxy-β-D-erythro-pentofuranosyl)-, Cordycepine, NSC 401022, NSC 63984, β-D-erythro-Pentofuranoside, adenine-9 3-deoxy-
IUPAC Name (2R,3R,5S)-2-(6-aminopurin-9-yl)-5-(hydroxymethyl)oxolan-3-ol
SMILES Nc1ncnc2c1ncn2[C@@H]3O[C@H](CO)C[C@H]3O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H13N5O3/c11-8-7-9(13-3-12-8)15(4-14-7)10-6(17)1-5(2-16)18-10/h3-6,10,16-17H,1-2H2,(H2,11,12,13)/t5-,6+,10+/m0/s1
Formula Weight 251.1018
Percent Purity >95%
Chemical Name or Material Cordycepin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.