missing translation for 'onlineSavingsMsg'
Learn More

(-)-Corey Lactone Diol, TRC

Product Code. 30349498
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30349498 1 g 1g
30356023 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30349498 Supplier Toronto Research Chemicals Supplier No. TRCC6949951G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32233-40-2
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
Synonym (3aR,4S,5R,6aS)Hexahydro-5-hydroxy-4-(hydroxymethyl)-2H-cyclopenta[b]furan-2-one, [3aR-(3aalpha,4alpha,5beta,6aalpha)]Hexahydro-5-hydroxy-4-(hydroxymethyl)-2H-cyclopenta[b]furan-2-one
IUPAC Name (3aR,4S,5R,6aS)-5-hydroxy-4-(hydroxymethyl)-3,3a,4,5,6,6a-hexahydrocyclopenta[b]furan-2-one
SMILES OC[C@H]1[C@H](O)C[C@@H]2OC(=O)C[C@H]12

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H12O4/c9-3-5-4-1-8(11)12-7(4)2-6(5)10/h4-7,9-10H,1-3H2/t4-,5-,6-,7+/m1/s1
Formula Weight 172.0736
Chemical Name or Material (-)-Corey Lactone Diol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.