Learn More
Tocris Bioscience™ CORM 3
Water-soluble carbon monoxide-releasing molecule
Brand: Tocris Bioscience™ 5320/50
This item is not returnable.
View return policy
Description
CORM 3 is a water-soluble carbon monoxide-releasing molecule. Suppresses thrombin-induced nitrite release in BV-2 microglia. Protects isolated rats hearts from reperfusion damage in vitro and is cardioprotective in a rat myocardial infarction model in vivo. Also prolongs survival of transplanted hearts in mice. Exhibits anti-inflammatory and vasorelaxant effects.Specifications
| CORM 3 | |
| 294.61 | |
| CORM 3 | |
| 50 mg |