missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 0.05% Trypsin/0.53mM EDTA in HBSS w/o Calcium, Magnesium or Sodium Bicarbonate
BIOPHARMA

Product Code. 15313651
Change view
Click to view available options
Quantity:
6 x 100 mL
6 x 500 mL
Unit Size:
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15313651 6 x 100 mL Pack of 6
15323651 6 x 500 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15313651 Supplier Corning™ Supplier No. 25052CI
  • ONLINE EXCLUSIVE PRICE
    81.12 €
    71.56 € / Pack of 6
    Save 9.56 €
    11% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used to enzymatically release adherent cells from tissue culture plates for passaging.

Trypsin is used to enzymatically release adherent cells from tissue culture plates for passaging.  Divalent cations such as calcium and magnesium, which are often present in the cell culture environment, inhibit this action.  EDTA sequesters these ions and thus enhances the efficacy of trypsin.  This product contains trypsin at a concentration of 0.05% with 0.53 mM EDTA, and does not contain sodium bicarbonate.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Form Liquid
Product Type Cell Culture Dissociation Reagent
Sterility Sterile
Without Additives No Calcium, No Magnesium, No Sodium Bicarbonate
pH 7 to 7.6
Quantity 6 x 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.