missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 0.05% Trypsin/0.53mM EDTA in HBSS w/o Calcium and Magnesium
BIOPHARMA

Product Code. 15303651
Change view
Click to view available options
Quantity:
6 x 100 mL
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15303651 6 x 100 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15303651 Supplier Corning™ Supplier No. 25051CI
  • ONLINE EXCLUSIVE PRICE
    81.60 €
    38.15 € / Pack of 6
    Save 43.45 €
    53% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used to enzymatically release adherent cells from tissue culture plates for passaging.

  • Contains trypsin at a concentration of 0.05% with 0.53mM EDTA, and sodium bicarbonate
  • Low endotoxin content
  • EDTA enhances the efficacy of trypsin
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Form Liquid
Product Type Cell Culture Dissociation Reagent
Sterility Sterile
Without Additives No Calcium, No Magnesium
pH 7 to 7.6
Quantity 6 x 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.