Learn More
Corning™ 384-Well Solid Black or White Polystyrene Microplates
Product Code. 15212239 Shop All Corning Products
Change view
Click to view available options
Quantity:
50/Cs.
Unit Size:
Pack of 50
Product Code. Quantity Unit Size
15212239 50/Cs. Pack of 50
1 options
Product Code. 15212239 Supplier Corning™ Supplier No. 3570BC
  • ONLINE EXCLUSIVE PRICE
    665.00 €
    617.09 € / Pack of 50
    Save 47.91 €
    7% Off
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Solid Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.
  • Minimal well to well crosstalk
  • White microplates enhance luminecent signals and have low background luminescence
  • Black mircoplates have low background fluorescence and minimize light scattering
  • Available untreated for solution-based assays, with high binding for immunoassay applications, or tissue culture treated for cell growth
  • Medical-grade polystyrene
  • Well volume: 112μL
  • Working volume: 20μL to 80μL
  • Surface area/well: 0.06cm2
  • Treated surface options are not coated, require no special handling and are stable at room temperature
  • Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
  • Plate dimensions meet proposed industry standards
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50/Cs.