Learn More
Corning™ 96-Well Clear Flat Bottom Sulfhydryl-BIND™ Microplate
For assays requiring site-directed orientation of a biomolecule, especially antibodies
Brand: Corning™ 2509
Code : 66
Additional Details : Weight : 0.02239kg
Description
- Cell culture microplates are sterile and nonpyrogenic.
- Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
Specifications
Standard | |
8 x 12 | |
Sulfhydryl-BIND | |
Polystyrene | |
75 to 200 μL | |
Sulfhydryl | |
Ideal for assays requiring site directed orientation of a biomolecule, especially antibodies | |
1/Pk. |
Without Lid | |
Individually Wrapped | |
Clear | |
96 | |
Non-sterile | |
Flat Bottom (F-well) | |
50/Cs. | |
50/Cs. |