missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well Microplate
Designed to reduce well-to-well crosstalk
Brand: Corning™ 3995
Code : 66
Additional Details : Weight : 1.72365kg
Description
Specifications
Standard | |
8 x 12 | |
White | |
Polystyrene | |
96 | |
Non-sterile | |
Flat Bottom (F-well) | |
25/Cs. | |
25/Cs. |
Without Lid | |
Bags of 5 | |
96-well Assay Microplate | |
360 μL | |
75 to 200 μL | |
Non-binding | |
Ideal for reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise | |
5/Pk. |