Learn More
Corning™ BioCoat™ Assay System
GREENER_CHOICE
Product Code. 10631932
Click to view available options
Type:
Cell Culture Insert System
Multi-well Insert System
Quantity:
1/Cs.
5/Cs.
Unit Size:
Each
Pack of 5
Product Code. 10631932
Brand: Corning™ 354802
Only null left
Add to basket
Add to basket
Includes
24-Multiwell™ Insert plate, specialized media to perform 24 assays, feeder tray, and lid.
Automation-friendly design
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
Corning BioCoat HTS Caco-2 Assay System, PET Membrane Contains specially formulated serum-free medium, culture supplements, sodium butyrate, and the Corning BioCoat Fibrillar Collagen 24-well insert system
TRUSTED_SUSTAINABILITY
Specifications
Closure Included Yes
Color Clear
Culture Area 0.3 cm2
Material PET
Sterility Non-sterile
Surface Treatment Collagen
Type Cell Culture Insert System
Well Count 24
For Use With (Application) Rapid differentiation of Caco 2 cells in vitro for robotic screening of prospective pharmaceuticals for oral bioavailability and absorption
Includes 24 multiwell insert plate, specialized media to perform 24 assays, feeder tray and lid
No. per Case 5/Ea.
No. per Pack 5/Pk.
Pore Size 1 μm
Quantity 5/Cs.