missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Collagen I Inserts with PET Membrane

Product Code. 10295091
Change view
Click to view available options
Pore Size:
0.3 μm
0.4 μm
0.45 μm
1 μm
3 μm
Type:
Cell Culture Insert
Well Plate
Quantity:
1
12/Pk., 24/Cs.
2
4
Unit Size:
Pack of 24
Pack of 4
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Type Quantity Unit Size
10295091 0.4 μm Well Plate 12/Pk., 24/Cs. Pack of 24
10133861 0.3 μm Cell Culture Insert 1 Pack of 4
10488262 1 μm Well Plate 2 Pack of 24
10366151 0.45 μm Well Plate 4 Pack of 24
10122851 3 μm Well Plate 4 Pack of 24
10526001 3 μm Well Plate 2 Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10295091 Supplier Corning™ Supplier No. 354444
  • ONLINE EXCLUSIVE PRICE
    550.00 €
    513.04 € / Pack of 24
    Save 36.96 €
    6% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines

  • Each lot is tested for the ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
  • Tested and found negative for bacteria and fungi
  • Cellware is also tested for ability to promote firm attachment of rat cerebellar granule cells
  • Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
  • Matrix components are applied as an even, optically clear coating
  • All lots tested and found negative for bacteria and fungi; cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells

For research use only.

Order Info

Bulk quantities are also available; contact your Fisher Scientific Customer Service Representative.

TRUSTED_SUSTAINABILITY

Specifications

Culture Area 0.3 cm2
Material PET
Pore Size 0.4 μm
Type Well Plate
For Use With (Application) Adhesion, growth and differentiation of neuronal and transfected cell lines
Quantity 12/Pk., 24/Cs.
Storage Requirements 2° to 8°C
Surface Coating Collagen
No. of Wells 24
Inserts per Plate 12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.