missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Collagen I Multiwell Plates
BIOPHARMAGREENER_CHOICE

Product Code. 10418662
Click to view available options
Culture Area:
9.6 cm2
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 10418662

Brand: Corning™ 356400

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Treated with BioCoat™ for enhanced cell attachment and growth of a variety of primary cells and ransformed cell lines in serum-free or reduced serum cultures. Corning™ BioCoat™ Collagen I Multiwell Plates closely mimmics in vivo environments for cell culture applications. Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines.

Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon, and bone. It is an integral part of the framework that holds cells and tissues together and has been recognized as a useful matrix for improving cell culture. in vitro use of collagen can exert effects on the adherence, morphology, growth, migration, and differentiation of a variety of cell types.

Applications include:

  • Promotion of cell attachment and spreading
  • Rapid expansion of cell populations
  • Serum-free or reduced serum culture
  • Cell adhesion assays
  • Improving survival of primary cells in culture

Source

  • Rat tail tendon
  • Quality Control

    • Tested for ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
    • Tested and found negative for bacteria and fungi
    • Collagen I purity >90% by SDS-PAGE

    Storage

  • 4°C to 30°C under dry conditions.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Array 2 x 3
    Certifications/Compliance ANSI, SBS
    Closure Color Clear
    Closure Included Yes
    Closure Material Polystyrene
    Bottom Shape Flat
    Color Clear
    Culture Area 9.6 cm2
    Material Polystyrene
    Shape Rectangular
    Sterility Non-sterile
    Surface Treatment Cell Culture
    Type Multi-well Cell Culture Plate
    Well Count 6
    For Use With (Application) Promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, cell adhesion assay, improving survival of primary cells in culture
    Includes Lid
    No. per Case 50/Cs.
    No. per Pack 5/Pk.
    Packaging 10 inner bags of 5 each
    Quantity 50/Cs.
    Source Rat Tail
    Storage Requirements 4°C to 30°C
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.