missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Collagen IV Multiwell Plates
BIOPHARMAGREENER_CHOICE

Product Code. 10469412
Change view
Click to view available options
Well Count:
6
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Well Count unitSize
10469412 6 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10469412 Supplier Corning™ Supplier No. 354428
Add to basket
Add to basket
This item is not returnable. View return policy

Type IV Collagen is a ubiquitous component in basement membranes and provides the major structural support for this matrix. Collagen IV is useful as a substrate for growth of epithelial, endothelial, muscle, and nerve cells.

  • Type IV Collagen is a ubiquitous component in basement membranes and provides the major structural support for this matrix. When the Collagen IV meshwork is assembled, it provides a scaffold for the assembly of other basement membrane components through interactions with laminin, entactin/nidogen, and heparan sulfate proteoglycan. Collagen IV is useful as a substrate for growth of epithelial, endothelial, muscle, and nerve cells. Collagen plays a role in the regulation of cell growth, differentiation and adhesion, as well as tissue formation.

Applications include:

  • Promotion of cell attachment and spreading
  • Cell differentiation and neurite outgrowth
  • Increased proliferation of PC12 cells
  • Studies of effects of collagen IV on cell behavior
  • Cell adhesion assays

Source

  • Engelbreth-Holm-Swarm (EHS) lathrytic mouse tumor

Quality Control

  • Tested for ability to initiate differentiation (neurite outgrowth) of NG-108 rat glioma/mouse neuroblastoma cells
  • Tested and found negative for bacteria and fungi
  • Collagen IV purity >90% by SDS-PAGE

Storage

  • 2°C to 8°C. Do not freeze.
TRUSTED_SUSTAINABILITY

Specifications

Array 2 x 3
Closure Color Clear
Closure Included Yes
Closure Material Polystyrene
Bottom Shape Flat
Color Clear
Material Polystyrene
Shape Rectangular
Sterility Non-sterile
Surface Treatment ECM/Attachment Factors
Type Multi-well Cell Culture Plate
Well Count 6
For Use With (Application) Promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, cell adhesion assay, improving survival of primary cells in culture
Includes Lid
No. per Case 5/Cs.
No. per Pack 1/Pk.
Quantity 5/Cs.
Source EHS Lathrytic Mouse Tumor
Storage Requirements 2°C to 8°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.