missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ BioCoat™ Laminin Multiwell Plate

Click to view available options
Culture Area:
9.6 cm2
Unit Size:
Pack of 5
Description
Laminin (LM), a major component of basement membranes, is a multifunctional glycoprotein that is used as a substrate to culture and maintain differentiated function of a wide variety of cells. Laminin has been shown in culture to stimulate neurite outgrowth, promote cell attachment, chemotaxis, cell differentiation, and neuronal survival.
Applications include:
- Promotion of cell attachment and spreading
- Induction of cell differentiation and neurite outgrowth
- Increases proliferation of myoblasts
- Studies of effects of laminin on cell behavior
- Cell adhesion assays
Source
Quality Control
- Tested for ability to initiate differentiation (neurite outgrowth) of NG-108 rat glioma/mouse neuroblastoma cells
- Tested and found negative for bacteria and fungi
- Laminin purity >90% by SDS-PAGE (contains entactin)
Storage
Specifications
Specifications
| Array | 2 x 3 |
| Closure Color | Clear |
| Closure Included | Yes |
| Closure Material | Polystyrene |
| Bottom Shape | Flat |
| Color | Clear |
| Culture Area | 9.6 cm2 |
| Material | Polystyrene |
| Shape | Rectangular |
| Sterility | Non-sterile |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction