Learn More
Corning™ BioCoat™ Matrigel™ Invasion Chamber with Corning™ Matrigel Matrix
Description
Corning BioCoat cell culture inserts are pre-coated with extracellular matrix proteins for applications requiring a protein-coated cell surface, such as cell adhesion, growth, invasion, migration and/or differentiation.
Coatings include Corning Matrigel™ matrix, Fibronectin, Collagen, and Laminin.
Specifications
Specifications
| Culture Area | 4.2 cm2 |
| Material | PET |
| Pore Size | 8 μm |
| For Use With (Application) | Studies in which minimal interference by growth factors is desirable |
| Quantity | 4 |
| No. per Pack | 6/Pk. |
| No. per Case | 24/Cs. |
| Surface Coating | Matrigel™ Matrix |
| Inserts per Plate | 6 |