missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Poly-L-Lysine Culture Dishes
GREENER_CHOICE

Product Code. 10666691
Click to view available options
Diameter (Metric):
35 mm
60 mm
No. per Pack:
20/pk.
5/pk
No. per Case:
100/cs.
20/cs.
Unit Size:
Pack of 100
Pack of 20
This item is not returnable. View return policy

Product Code. 10666691

Brand: Corning™ 354517

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provide excellent substrate for cell attachment and differentiation of primary neurons, glial, neuroblastoma and glioma cells

Suitable for promoting adhesion of transfected cell lines and other fastidious cell types. Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology.

  • Minimize assay failure due to cell detachment
  • Matrix components are applied as an even, optically clear coating
  • Precoated with Poly-L-Lysine to eliminate time-consuming preparation
  • Lot-to-lot consistency provides reproducible experiments
  • All lots tested and found negative for bacteria and fungi
  • Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells

For adhesion, growth and differentiation of neuronal and transfected cell lines

Order Info

Bulk quantities available; please contact your Customer Service Representative.

TRUSTED_SUSTAINABILITY

Specifications

Bottom Shape Round
Culture Area 21.3 sq. cm
Shape Round
Surface Treatment Poly-L-Lysine
Type Cell Culture Dish
No. per Case 20/cs.
No. per Pack 5/pk
Diameter (Metric) 60 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.