missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Sterile Cell Strainers
BIOPHARMA

Product Code. p-4830339 Shop All Corning Products
Click to view available options
Mesh Size:
40 μm
70 μm
100 μm
Color:
Blue
White
Yellow
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 15360801

Brand: Corning™ 431750

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Isolates primary cells rapidly to consistently obtain a uniform single-cell suspension from tissues. Corning™ Sterile Cell Strainers are ideal for preparation of flow cytometry samples and are used with stem cells and primary cells.

 

  • Strong nylon mesh available in three pore sizes (40, 70 or 100 μm) for variety of applications
  • Fits all major brands of 50 mL conical tubes
  • Non cytotoxic device has color-coded polypropylene frames
  • Grip area for handling with forceps
  • Sterilized by gamma irradiation
  • Use for pre-filtering solutions containing particulates >40μm and for cleaning cell suspensions prior to sub culturing, counting and analysis or freezing
  • Individually packed in easy to open bags

 

For preparation of:

  • Single cell suspensions of blood cells
  • Specimens for primary cell cultures and immunogens
  • Primary cells from tissue

 

TRUSTED_SUSTAINABILITY

Specifications

Mesh Material Nylon
Color Blue
Material Polypropylene
Sterility Sterile
For Use With (Equipment) Conical Tubes, 50 mL
Mesh Size 40 μm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.