missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Cell-Tak Cell and Tissue Adhesive
BIOPHARMA

Product Code. 10317081 Shop All Corning Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10317081 1 mg 1mg
11593550 10 mg 10mg
11583550 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10317081 Supplier Corning™ Supplier No. 354240
  • ONLINE EXCLUSIVE PRICE
    334.00 €
    312.03 € / 1mg
    Save 21.97 €
    6% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Attach cells or tissue sections to many types of surfaces, including plastic, glass, metal, FEP Polymer, and biological materials. Corning™ Cell-Tak Cell and Tissue Adhesive is a specially formulated protein solution extracted from marine mussel, Mytilus edulis.

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Stable for at least three months at 2° to 8°C. Do not freeze.
Description Molecular weight: 110 to 114kd
Concentration 1 to 5μg/ cm2 of growth surface depending on cell type
Form Liquid
Product Type Cell and Tissue Adhesive
Formulation As liquid in 5% acetic acid
Quantity 1 mg
Source Polyphenolic Proteins secreted by Mytilus edulis
compliance-icons
Product Identifier
  • Cell-Tak Cell and Tissue Adhesive
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362-Take off contaminated clothing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.