missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CellBIND™ Surface Microplates
BIOPHARMAGREENER_CHOICE

Product Code. 10739864 Shop All Corning Products
Click to view available options
Culture Area:
0.95 cm2
1.9 cm2
3.8 cm2
9.5 cm2
Capacity (Metric):
0.28 mL (Well)
0.57 mL (Well)
2.9 mL (Well)
6.9 mL
Volume (Metric) Working:
0.19 to 0.28 mL
0.38 to 0.57 mL
0.76 to 1.14 mL
1.9 to 2.9 mL
Well Count:
6
12
24
48
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 10739864

Brand: Corning™ 3336

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Treated with CellBIND™ to help cells adapt to reduced serum or serum-free media and improve cell attachment for increased cell growth and yield. Corning™ CellBIND™ Surface Microplates are constructed of clear polystyrene for optimum visibility and are available in multiple well formats.

  • Individual alphanumerical codes for well identification, flat bottoms
  • Treated for optimal cell attachment (except where noted)
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
  • Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
  • Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
  • Sterile
  • Nonpyrogenic

Certifications

Certified nonpyrogenic

TRUSTED_SUSTAINABILITY

Specifications

Closure Included Yes
Capacity (Metric) 6.9 mL
Culture Area 3.8 cm2
Material Polystyrene
Shape Round
Sterility Sterile
Surface Treatment CellBIND
Type Cell Culture Plate
Volume (Metric) Working 0.76 to 1.14 mL
Well Count 12
No. per Case 50/Cs.
No. per Pack 5/Pk.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.