Learn More
Corning™ CellCube™ Tubing Adaptor
Shop All Corning Products
Click to view available options
Product Type:
4 AseptiQuik™ S Connectors, 4 Tubes Clamps
AseptiQuik™ S Connected to a Female MPC
AseptiQuik™ S Connected to a Male MPC
Unit Size:
Pack of 4
Description
- Closed System Cell Cube
- CellCube 10, 25, 100 modules, circulation loops, and connectors
- Allows for easy adherent cell scale-up that integrate seamlessly with AseptiQuik™ and MPC connectors.
Specifications
Specifications
| Diameter (Metric) Inner Tubing | 9.53 mm |
| Includes | Thermoelastic Tubing, Aseptic Connector to Male MPC |
| No. per Case | 4/Cs. |
| No. per Pack | 1/Pk. |
| Product Type | AseptiQuik™ S Connected to a Male MPC |