Learn More
Corning™ CellCube™ Tubing Manifold
Shop All Corning Products
Click to view available options
:
2/Cs.
Description
- Closed System Cell Cube
- CellCube 10, 25, 100 modules, circulation loops, and connectors
- Allows for easy adherent cell scale-up that integrate seamlessly with AseptiQuik™ and MPC connectors.
Specifications
Specifications
| Includes | Thermoelastic Tubing, Cross Connection, 4 Aseptic Connectors |
| No. per Case | 2/Cs. |
| No. per Pack | 1/Pk. |