missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Cell Culture Buffers: Dulbecco's Phosphate-Buffered Salt Solution 10X

Click to view available options
With Additives:
None, Magnesium
Unit Size:
Pack of 6
Description
Dulbecco's Phosphate Buffered Saline (DPBS) is a buffer solution which can be used to maintain cell culture media in the physiological pH range of 7.0 to 7.6. It can serve as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time.
Specifications
Specifications
| Content And Storage | 2° to 30°C |
| Concentration | 10 X |
| Form | Liquid |
| Product Type | Dulbecco's Phosphate Buffered Saline |
| With Additives | None, Magnesium |
| Formulation | Calcium Chloride 1g/L, Potassium Chloride 2g/L, Potassium Dihydrogen Phosphate 2g/L, Magnesium Chloride Hexahydrate 1g/L, Sodium Chloride 80g/L, Disodium Phosphate 11.50g/L |
| pH | 4.1 to 4.7 |
| Quantity | 6 x 500 mL |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion