missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Microbiological Media
BIOPHARMA

Product Code. 15343721
Change view
Click to view available options
Media Type:
LB Broth
Terrific Broth
Unit Size:
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Media Type Unit Size
15343721 Terrific Broth Pack of 6
15333721 LB Broth Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15343721 Supplier Corning™ Supplier No. 46055CM
  • ONLINE EXCLUSIVE PRICE
    320.00 €
    104.08 € / Pack of 6
    Save 215.92 €
    67% Off
 Restricted Item
Estimated Shipment: 19-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sterile filtered

  • For the maintenance and propagation of E. coli
  • Terrific Broth supports higher cell densities than LB Broth, increasing plasmid yield

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Manufacturing Origin United States
Media Type Terrific Broth
Fill Volume 6 x 1000 mL
Formulation Casein Peptone 10g/L, NaCl 10g/L
For Use With (Application) For maintenance/propagation of Escherichia coli with plasmid DNA and recombinant Protein production
Packaging Type Bottle
Quantity 6 x 1000 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.