Learn More
Reagent Grade Water Tested to USP Purified Water, Corning™
BIOPHARMA
Product Code. 15313711
Change view
Click to view available options
Quantity:
2 x 4 L
Unit Size:
Pack of 2
Product Code. Quantity Unit Size
15313711 2 x 4 L Pack of 2
1 options
Product Code. 15313711 Supplier Corning Media Tech Supplier No. 46002LF
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 05-04-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Used in common laboratory applications requiring highly pure water. Corning™ Reagent Grade Water Tested to USP Purified Water is used in the preparation of reagents, rinsing glassware and plastic ware.
  • Tested to USP monograph for purified water
  • DNase, RNase, Protease-free
  • Endotoxin levels ≤0.025EU/mL
Certifications
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2 x 4 L