missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Color-Coded Cap Inserts
GREENER_CHOICE

Product Code. 10474912 Shop All Corning Products
Change view
Click to view available options
Closure Color:
Assorted
Blue
Green
Red
White
Yellow
Unit Size:
Pack of 500
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Color unitSize
10474912 Blue Pack of 500
10317532 White Pack of 500
10664933 Yellow Pack of 500
10032202 Green Pack of 500
10377572 Red Pack of 500
10461822 Assorted Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10474912 Supplier Corning™ Supplier No. 2016
Add to basket
Add to basket
This item is not returnable. View return policy

For use with Corning cryogenic vials except 03-374-6

  • Cap inserts provide color coding for easy sample identification.
  • Inserts are packaged in resealable bags.
  • Nonsterile
  • Cap inserts fit all Corning™ cryogenic vials.
TRUSTED_SUSTAINABILITY

Specifications

Closure Color Blue
Color Blue
Material Polypropylene
Sterility Non-sterile
No. per Case 500
No. per Pack 50
Closure Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.