Learn More
Corning™ Costar™ Transtar-96™ 96-Well Liquid Transfer System
Product Code. 10471052 Shop All Corning Products
Change view
Click to view available options
Channels:
96
Unit Size:
Each
Product Code. Channels unitSize
10471052 96 Each
1 options
Product Code. 10471052 Supplier Corning™ Supplier No. 7605
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Portable, autoclavable liquid handling system designed for use with 96 well plates.
The Costar™ Transtar-96™ System is a portable, autoclavable liquid handling system designed for use with 96 well plates. A sterile 96 tip disposable cartridge, which loads into the Transtar-96™ system, enables liquids to be aspirated, transferred and dispensed over a volume range of 25 to 200 μL in 5 μL increments. -Ideal for changing cell culture media and screening monoclonal antibodies. -Accuracy is rated at +/- 5% at all volume levels. -For sterile replacement cartridges, see No. 7610. -For sterile replacement reservoirs, see Nos. 4876 (open), 4877 (12-channel), or 4878 (8-channel). Refer to accessories tab below.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Liquid Transfer System
Autoclavable Autoclavable
Channels 96
For Use With (Application) Designed for use with 96-well plates, changing cell culture media and screening monoclonal antibodies
Sterility Sterile