Learn More
Corning™ Untreated Culture Dishes
Binds bio-molecules through passive interactions and hydrophobic in nature. Corning™ Untreated Culture Dishes provide an ideal environment for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface.
194.74 € - 401.64 €
Specifications
Closure Color | Clear |
---|---|
Closure Included | Yes |
Closure Material | Polystyrene |
Bottom Shape | Flat |
Color | Clear |
Product Code | Brand | Culture Area | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Culture Area | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | |||
10429141
|
Corning™
430597 |
148 cm2 | 150 mm | 25 mm |
194.74 €
Pack of 60 |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10609321
|
Corning™
430588 |
8 cm2 | 35 mm | 10 mm |
232.90 €
Pack of 500 |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10419141
|
Corning™
430591 |
55 cm2 | 100 mm | 20 mm |
401.64 €
Pack of 500 |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||
10409141
|
Corning™
430589 |
21 cm2 | 60 mm | 15 mm |
239.39 €
Pack of 500 |
Estimated Shipment: 09-09-2024
Log in to see stock. |
|||
Description
- Manufactured from optically clear virgin polystyrene
- Not treated for applications where cell attachment is not desired
- Not treated for applications where cell attachment is not desired
- Stacking beads aid in handling
- Vents provide consistent gas exchange
- Sterile
- Sterile
- Nonpyrogenic
Dish style (mm) = actual growth surface diameters: 35 mm dish = 34.4 mm; 60 mm dish = 52.1 mm; 100 mm dish = 83.8 mm; 150 mm dish = 139.1 mm.
Cat. no. 431111, Square dishes with interior bottom dimensions of 224 x 224 mm.
Order Info
For Cell Culture Treated Polystyrene Dishes: Shelf pack 430293 consists of six-pack carriers which hold six sleeves of 10 dishes each–minimizing contamination of unused dishes after sleeve is opened. Supplied in sleeves of 20 dishes (except 150 x 25mm dish which is packaged five per sleeve and 245 x 245mm dish which comes four per sleeve).
Specifications
Clear | |
Polystyrene | |
Clear | |
Polystyrene | |
Sterile | |
Cell Culture Petri Dish |
Yes | |
Flat | |
Single-use | |
Round | |
Non-treated | |
Lid |