missing translation for 'onlineSavingsMsg'
Learn More

Corning™ DeckWorks™ Bulk Pipet Tips

Product Code. 11901841 Shop All Corning Products
Click to view available options
Volume (Metric):
0.1 to 10 μL
1 to 200 μL
100 to 2000 μL
Color:
Natural
Yellow
Unit Size:
Pack of 1000
Pack of 10000
Pack of 768
Pack of 7680
This item is not returnable. View return policy

Product Code. 11901841

Brand: Corning™ 4110

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Compatible with DeckWorks racks and packaged in resealable polyethylene bags

Corning DeckWorks bulk pipet tips are compatible with DeckWorks racks and packaged in resalable polyethylene bags. Tip sizes include 10 μL, 200 μL, and 1000 μL and offer ­maximum savings.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.1 to 10 μL
Sterility Non-sterile
Color Natural
For Use With (Application) Help reduce the production and waste of plastic materials
Tip Style Round
Packaging Bulk
Disposable Single-use
For Use With (Equipment) DeckWorks Racks
Graduated Yes
Product Line DeckWorks
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.