missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ DeckWorks™ Hinged Rack Barrier Tips
Shop All Corning Products
Click to view available options
Volume (Metric):
100 to 1000 μL
Unit Size:
Pack of 768
Beskrivning
- The hinged rack design allows easy rack stacking
- Racks can be reused
- Simply remove the tip insert in the rack and replace with the Corning tip inserts for hinged racks
- Allow maximize storage space and help reduce the production and waste of plastic materials
- Hinged racks within this system are manufactured from recycled polypropylene
- Used for quantitative assays in which sample binding can significantly alter results
- These sterile tips will not trap liquids or inhibit PCR in the event of over-pipetting
Specifikationer
Specifikationer
| Volume (Metric) | 100 to 1000 μL |
| Filtered | Filtered |
| Sterility | Sterile |
| Color | Natural |
| Clean Claims | Nonpyrogenic, Human DNA-free, RNase-/DNase-free, Sterility Assurance Level 10-3 |
| For Use With (Application) | Used for quantitative assays in which sample binding can significantly alter results |
| Format | Hinged Rack |
| Tip Style | Round |
| Packaging | 96 Tips/Rack, 8 Racks/Pack |
| Graduated | Yes |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering