missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ DeckWorks™ Low Binding Hinged Rack Barrier Tips
Shop All Corning Products
Click to view available options
Volume (Metric):
0.1 to 10 μL
1 to 100 μL
1 to 20 μL
1 to 200 μL
Unit Size:
Pack of 3840
Pack of 960
Description
Corning DeckWorks low binding barrier tips feature an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures. DeckWorks low binding barrier tips will not trap liquids or inhibit PCR in the event of over pipetting.
Specifications
Specifications
| Volume (Metric) | 1 to 200 μL |
| Filtered | Filtered |
| Sterility | Sterile |
| Color | Natural |
| For Use With (Application) | Used for quantitative assays in which sample binding can significantly alter results |
| Format | Hinged Rack |
| Tip Style | Low Retention, Round |
| Packaging | Rack |
| Graduated | Yes |
| Product Line | DeckWorks |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction