missing translation for 'onlineSavingsMsg'
Learn More

Corning™ DeckWorks Pipet Tips, Nonsterile

Product Code. 11951841 Shop All Corning Products
Click to view available options
Volume (Metric):
0.1 to 10 μL
1 to 200 μL
1 to 300 μL
100 to 1000 μL
Color:
Natural
Yellow
Unit Size:
Pack of 3072
Pack of 3840
Pack of 768
Pack of 960
This item is not returnable. View return policy

Product Code. 11951841

Brand: Corning™ 4116

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Universal tip fits in hinged DeckWorks racks that allows for easy rack stacking

Corning universal fit tips in hinged DeckWorks racks are available in 10 μL, 200 μL, 300 μL, and 1,000 μL sizes. The 10 μL, 300 μL, and 1,000 μL tips are extended length and allow for improved access into microcentrifuge (1.5 mL) and standard conical bottom (15/50 mL) centrifuge tubes. Robust 96-tip racks are designed for repeated steam autoclave cycles and can be reused by removing the empty grey tip deck and replacing it with a new reload deck of 96 tips from the DeckWorks reload system. Corning racked 300 μL DeckWorks pipet tips are designed for all popular 300 μL multi-channel instruments.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 200 μL
Sterility Non-sterile
Color Natural
For Use With (Application) Universal tip fits in hinged DeckWorks racks that allows for easy rack stacking
Format Hinged Rack
Tip Style Round
Packaging Hinged rack
Product Line DeckWorks
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.