missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Dispase

Product Code. 11553550 Shop All Corning Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11553550 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11553550 Supplier Corning™ Supplier No. 354235

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bacillus-derived neutral metalloprotease that is recommended for recovering cells cultured on BD Matrigel™ Basement Membrane Matrix

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Recovering cells cultured on BD Matrigel™ basement membrane matrix
Content And Storage -20°C
Source Bacillus polymyxa Metallo Protease
Quantity 100 mL
Product Type Dispase
Molecular Weight (g/mol) 36
compliance-icons compliance-icons
Product Identifier
  • Dispase
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.