missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Donor Calf Serum
Descripción
- Used as a supplement to cell culture media to help sustain growth and metabolism
- Each batch of raw serum is screened for endotoxins, residual hemoglobin concentration, pH and microbiological quality before being used in production
- Each lot is 0.1μm triple-filtered, low in endotoxins and final tested for the absence of mycoplasma and viruses
For research use only
Compliance
Sera is aseptically collected and processed in a facility that is compliant with FDA guidelines for Class I Medical Devices.
Especificaciones
Especificaciones
| Certifications/Compliance | FDA guidelines for Class I Medical Devices |
| Species | Bovine |
| Product Type | Calf Bovine Serum |
| Quantity | 500 mL |
| For Use With (Application) | Research use, used as a supplement to cell culture media to help sustain growth and metabolism |
| Mycoplasma Detected | No |
| USDA Safety Testing Included | Yes |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?