missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Fibronectin, Human

Product Code. 10526961 Shop All Corning Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10526961 1 mg 1mg
11533610 5 mg 5mg
11543610 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10526961 Supplier Corning™ Supplier No. 354008
  • ONLINE EXCLUSIVE PRICE
    171.00 €
    159.46 € / 1mg
    Save 11.54 €
    6% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Promotes cellular attachment, spreading, and proliferation on tissue-culture surfaces. Corning™ Fibronectin, Human can be used as an additive to serum-free culture media.

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

The principal functions of fibronectin appear to be in cellular migration during wound healing and development, regulation of cell growth and differentiation, and haemostasis/thrombosis.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 8°C
Product Type Fibronectin
pH 11
Quantity 1 mg
Source Human Plasma
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.