missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Flexible Packaging Systems: MPC Connector Tubing

Product Code. 31932499 Shop All Corning Products
Change view
Click to view available options
Tubing Style:
Fits tubing 60.96 cm
Fits tubing 91.44 cm
Unit Size:
Each
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tubing Style unitSize
31932499 Fits tubing 91.44 cm Pack of 10
17243594 Fits tubing 91.44 cm Each
17253594 Fits tubing 60.96 cm Each
31932500 Fits tubing 60.96 cm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31932499 Supplier Corning™ Supplier No. 9170004
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
Denna artikel kan inte returneras. Se returpolicy

Clear tubing; female MPC connector

  • Chemically resistant
  • Heat sealable
  • Flexible

TRUSTED_SUSTAINABILITY

Specifikationer

Content And Storage Male luer Cap
Tubing Style Fits tubing 91.44 cm
Product Type Connector tubing
Certifications/Compliance cGMPs, FDA Regulation 21 CFR part 820 and ISO 13485 Quality
Color Clear
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.