missing translation for 'onlineSavingsMsg'
Learn More

Corning™ GL32 Orange Polypropylene Screw
GREENER_CHOICE

Product Code. 17273474 Shop All Corning Products
Change view
Click to view available options
Thread Size (Metric):
GL 32
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size (Metric) unitSize
17273474 GL 32 Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17273474 Supplier Corning™ Supplier No. 139532LTC
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use a linerless plug seal design for efficient sealing

These are various sizes and types of screw caps for use with storage bottles.

TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Material Polypropylene
No. per Case 20/Cs.
No. per Pack 20/Pk.
Sterility Non-sterile
Thread Size (Metric) GL 32
Neck Finish 39 mm
Product Type Plug Seal
For Use With (Equipment) Storage bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.